Nonyl-β-D-glucopyranosid
Lieferant: BIOMOL RESEARCH LABORATORIES
Synonyme: Nonyl-β-D-glucosid, NG
50862-1 50862-2
BMOL50862-1EA 0 EUR
BMOL50862-1 BMOL50862-2
Nonyl-β-D-glucopyranosid
Nonyl-β-D-glucopyranosid
Formel: C₁₅H₃₀O₆
Molecular Weight: 306,4 g/mol
MDL: MFCD00063300
CAS-Nummer: 69984-73-2