N-Octyl-β-D-glucopyranosid
Lieferant: BACHEM BIOCHEMICA
|
Achtung
|
Synonyme:
Octyl-β-D-glucosid, Octyl-β-D-glucopyranosid
4005687.0005
4005687.0025
4005687.0001
BACE4005687.0005EA
0
EUR
BACE4005687.0005
BACE4005687.0025
BACE4005687.0001
N-Octyl-β-D-glucopyranosid
N-Octyl-β-D-glucopyranosid
Formel:
C₁₄H₂₈O₆ Molecular Weight: 292.37 g/mol Schmelzpunkt: 110 Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063288 CAS-Nummer: 29836-26-8 EINECS: 249-887-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...