N-Octyl-β-D-glucopyranosid, Millipore®
Lieferant: Merck Millipore Calbiochem
Achtung
Synonyme: Octyl-β-D-glucosid, Octyl-β-D-glucopyranosid
494460-1 494460-5
494460-1EA 385 EUR
494460-1 494460-5
N-Octyl-β-D-glucopyranosid, Millipore®
N-Octyl-β-D-glucopyranosid
Formel: C₁₄H₂₈O₆
Molecular Weight: 292,37 g/mol
Schmelzpunkt: 110 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063288
CAS-Nummer: 29836-26-8
EINECS: 249-887-8