di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP) 20 mg, Tabletten, Sigma-Aldrich®
Lieferant: Merck
Synonyme: pNPP, ortho-Phosphorsäure(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, 4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, p-Nitrophenylphosphat Dinatriumsalz Hexahydrat, ortho-Phosphorsäuremono(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, Phosphatasesubstrat, 4-Nitrophenyldihydrogenphosphat Dinatriumsalz Hexahydrat
M-Clarity™ Qualitätsstufe = MQ300
N2765-100TAB N2765-50TAB
SIALN2765-100TABEA 665 EUR
SIALN2765-100TAB SIALN2765-50TAB
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP) 20 mg, Tabletten, Sigma-Aldrich®
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)
Chromogenic substrate for the determination of acid and alkaline phosphatases.
The reaction produces a soluble end product that is yellow in color. The reaction may be stopped with 3 N NaOH and read at 405 nm
Formel: C₆H₄NNa₂O₆P·6H₂O
Molecular Weight: 371.144 g/mol
Schmelzpunkt: 300
Lagertemperatur: Gefrierschrank
MDL: MFCD00066288
CAS-Nummer: 333338-18-4
EINECS: 224-246-5