15 mg, Tabletten, Sigma-Aldrich®
Lieferant: Merck
Synonyme: pNPP, ortho-Phosphorsäure(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, 4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, p-Nitrophenylphosphat Dinatriumsalz Hexahydrat, ortho-Phosphorsäuremono(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, Phosphatasesubstrat, 4-Nitrophenyldihydrogenphosphat Dinatriumsalz Hexahydrat
M-Clarity™ Qualitätsstufe = MQ200
N2640-100TAB N2640-50TAB
SIALN2640-100TABEA 643 EUR
SIALN2640-100TAB SIALN2640-50TAB
15 mg, Tabletten, Sigma-Aldrich®
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)
4-Nitrophenyl phosphate disodium salt hexahydrate has been used in enzyme-linked immunosorbent assay (ELISA) as a substrate for streptavidin-Alkaline Phosphatase. It has also been used as a substrate in control assay for comparing 4-aminophenyl phosphate/diethanolamine (APP/DEA) assay for alkaline phosphatase.
Formel: C₆H₄NNa₂O₆P·6H₂O
Molecular Weight: 371.144 g/mol
Schmelzpunkt: 300
Lagertemperatur: Gefrierschrank
MDL: MFCD00066288
CAS-Nummer: 333338-18-4
EINECS: 224-246-5