Strontiumaluminat ≥99,5% (Metall-Basis)
Lieferant: Thermo Scientific
39647.14
39647.22
39647.14EA
114
EUR
39647.14
39647.22
Strontiumaluminat ≥99,5% (Metall-Basis)
Strontium aluminate
Purity: 99.5%
CAS No.: 12004-37-4
Molecular Formula: SrAl2O4
Formula Weight: 205.58
Physical Form: -100 Mesh Powder
Density: 3.559
Appearance: Silver
Odour: Odourless
MDL No.: MFCD00054054
Formel:
Al₂O₄Sr Dichte: 3,559 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054054 CAS-Nummer: 12004-37-4 EINECS: 234-455-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...