Vanadylsulfat Hydrat ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: basisches Vanadiumsulfat Hydrat, Vanadium(IV)oxidsulfat Hydrat, Vanadiumoxidsulfat Hydrat, Vanadium(IV)sulfatoxid Hydrat
11090.18 11090.30 11090.09 AB202309.0050
11090.18EA 120 EUR
11090.18 11090.30 11090.09 ABCRAB202309.0050
Vanadylsulfat Hydrat ≥99,9% (Metall-Basis)
Vanadylsulfat Hydrat
Purity: 99.9% CAS No.: 123334-20-3 Molecular Formula: VOSO4.xH2O Formula Weight: 163.00(anhy) Physical Form: Lump Density: 2.5 Appearance: Blue Odour: Odourless Solubility: Soluble in water Merck Reference: 14,9926 UN No.: 2931 MDL No.: MFCD00149933
Formel: O₅SV·H₂O
Molecular Weight: 163,00 g/mol (anhy)
Dichte: 2,5
Lagertemperatur: Raumtemperatur
MDL: MFCD00149933
CAS-Nummer: 123334-20-3
EINECS: 248-652-7
UN: 2931
ADR: 6.1,II
Merck Index: 14,9926