3-Indoleacrylic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

122180010 122180050
122180010.EA 59.2 EUR
122180010. 122180050.
3-Indoleacrylic acid 98%
3-Indoleacrylic acid
Formel: C₁₁H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00005633
CAS-Nummer: 1204-06-4
EINECS: 214-872-7

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to beige powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 180°C to 190°C
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR