Bis(4-nitrophenyl)disulfid 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,2-Bis(4-nitrophenyl)disulphane, 4-Nitrophenyl disulphide, 4,4'-Dinitrodiphenyl Disulphide

128790050 128790250
128790050.EA 23.9 EUR
128790050. 128790250.
Bis(4-nitrophenyl)disulfid 97%
Bis(4-nitrophenyl)disulfid
Formel: C₁₂H₈N₂O₄S₂
Siedepunkt: 478,8°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003573
CAS-Nummer: 100-32-3
EINECS: 202-840-5

Bestellen

Spezifikation Testergebnisse

Appearance light yellow to yellow to orange powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %
Melting point 181°C to 186°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR