4H-Pyran-4-on 98%

Lieferant: Thermo Scientific

Synonyme: gamma-Pyrone

175750010 175750050
175750010.EA 161 EUR
175750010. 175750050.
4H-Pyran-4-on 98%
4H-Pyran-4-on
Formel: C₅H₄O₂
Molecular Weight: 96,09 g/mol
Siedepunkt: 210…215°C (760mmHg)
Flammpunkt: 101°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006576
CAS-Nummer: 108-97-4
EINECS: 203-634-8

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow to brown low melting solid
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 32°C to 34°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR