5-Phenyl-1,3-oxazole 97%
Lieferant: Thermo Fisher Scientific
264220010
264220050
264220010.EA
83.2
EUR
264220010.
264220050.
5-Phenyl-1,3-oxazole 97%
5-Phenyl-1,3-oxazole
Formel:
C9H7NO Siedepunkt: 261°C (760mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067074 CAS-Nummer: 1006-68-4 |
Spezifikation Testergebnisse
Appearance | yellow to beige to brown low melting mass or crystals |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...