Glycocholic acid hydrate

Lieferant: Thermo Fisher Scientific

466790010 466790050
466790010.EA 51.2 EUR
466790010. 466790050.
Glycocholic acid hydrate
Glycocholic acid hydrate
Formel: C₂₆H₄₃NO₆·xH₂O
Molecular Weight: 465.63 g/mol
MDL: MFCD06408004
CAS-Nummer: 1192657-83-2

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Water ≤8 %
Solubility (50 mg/ml in ethanol) Clear colorless to light yellow solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR