Amprenavir

Lieferant: Thermo Fisher Scientific

466980250 466981000
466980250.EA 88.5 EUR
466980250. 466981000.
Amprenavir
Amprenavir
Formel: C₂₅H₃₅N₃O₆S
Lagertemperatur: Gefrierschrank
MDL: MFCD00934214
CAS-Nummer: 161814-49-9

Bestellen

Spezifikation Testergebnisse

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with HClO4 96.0 to 104.0 % (on dry substance)
Water ≤7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR