5-Methyl-2-pyrazincarbonsäure-4-oxid 98+%

Lieferant: Thermo Scientific
Achtung

Synonyme: Acipimox, 5-Carboxy-2-methylpyrazine 1-oxide, 2-Carboxy-5-methylpyrazine 4-Oxide, 5-Carboxy-2-methylpyrazine 1-Oxide

466990010 466990050
466990010.EA 65.4 EUR
466990010. 466990050.
5-Methyl-2-pyrazincarbonsäure-4-oxid 98+%
5-Methyl-2-pyrazincarbonsäure-4-oxid
Formel: C₆H₆N₂O₃
Molecular Weight: 154,13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00865757
CAS-Nummer: 51037-30-0
EINECS: 256-928-3
Merck Index: 13,00113

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥98.0 %
Melting point 180°C to 190°C
Loss on drying ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR