Rilpivirine

Lieferant: Thermo Scientific
Achtung

Synonyme: (E)-4-((4-((4-(2-Cyanovinyl)-2,6-dimethylphenyl)amino)pyrimidin-2-yl)amino)benzonitrile

467720010 467722500
467720010.EA 744 EUR
467720010. 467722500.
Rilpivirine
Rilpivirine
Formel: C₂₂H₁₈N₆
Molecular Weight: 366,42 g/mol
Siedepunkt: 634°C (760mmHg)
Lagertemperatur: Gefrierschrank
MDL: MFCD11046372
CAS-Nummer: 500287-72-9

Bestellen

Spezifikation Testergebnisse

Appearance white to beige to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR