Andrographolide

Lieferant: Thermo Scientific

Synonyme: (S,E)-4-Hydroxy-3-(2-((1R,4aS,5R,6R,8aS)-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylenedecahydronaphthalen-1-yl)ethylidene)dihydrofuran-2(3H)-one

468290010 468290050
468290010.EA 53.7 EUR
468290010. 468290050.
Andrographolide
Andrographolide
Formel: C₂₀H₃₀O₅
Molecular Weight: 350,45 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07778082
CAS-Nummer: 5508-58-7
EINECS: 226-852-5
Merck Index: 13,00638

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 229°C to 235°C
Specific optical rotation -124° to -129° (20°C, 589 nm) (1.5% in acetic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR