Fidaxomicin

Lieferant: Thermo Scientific

Synonyme: Lipiarmycin A3, Tiacumicin B, (2R,3S,4S,5S,6R)-6-(((3E,5E,8S,9E,11S,12R,13E,15E,18S)-12-(((2R,3S,4R,5S)-3,4-Dihydroxy-5-(isobutyryloxy)-6,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-11-ethyl-8-hydroxy-18-((R)-1-hydroxyethyl)-9,13,15-trimethyl-2-oxooxacyclooctadeca-3,5,9,13,15-pentaen-3-yl)methoxy)-4-hydroxy-5-methoxy-2-methyltetrahydro-2H-pyran-3-yl 3,5-dichloro-2-ethyl-4,6-dihydroxybenzoate, Clostomicin B1

468310010 468312500
468310010.EA 833 EUR
468310010. 468312500.
Fidaxomicin
Fidaxomicin
Formel: C₅₂H₇₄Cl₂O₁₈
Molecular Weight: 1058,04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD27976367
CAS-Nummer: 873857-62-6

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 160°C to 170°C
Water ≤3 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR