Cefpiramide

Lieferant: Thermo Scientific

Synonyme: (6R,7R)-7-((R)-2-(4-hydroxy-6-methylnicotinamido)-2-(4-hydroxyphenyl)acetamido)-3-(((1-methyl-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

468331000 468335000
468331000.EA 94.5 EUR
468331000. 468335000.
Cefpiramide
Cefpiramide
Formel: C₂₅H₂₄N₈O₇S₂
Molecular Weight: 612,63 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00864893
CAS-Nummer: 70797-11-4

Bestellen

Spezifikation Testergebnisse

Appearance white to off-white powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Water ≤9 %
Specific optical rotation -100° to -112° (20°C, 589 nm) (10 mg/ml in DMF) (on dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR