Benzo[cd]indol-2(1H)-on

Lieferant: Thermo Scientific
Achtung

Synonyme: Naphthostyril, Naphtholactam, Benz[cd]indol-2(1H)-one

468400010 468400050 468400250
468400010.EA 35 EUR
468400010. 468400050. 468400250.
Benzo[cd]indol-2(1H)-on
Benzo[cd]indol-2(1H)-on
Formel: C₁₁H₇NO
Molecular Weight: 169,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009748
CAS-Nummer: 130-00-7
EINECS: 204-973-4

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to brown powder or crystals
Infrared spectrum Conforms
HPLC ≥88 %
Melting point 179°C to 183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR