Nizatidin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine, N-(2-(((2-((Dimethylamino)methyl)thiazol-4-yl)methyl)thio)ethyl)-N-methyl-2-nitroethene-1,1-diamine
468470050
468470250
468470050.EA
82.1
EUR
468470050.
468470250.
Nizatidin
Nizatidin
Formel:
C₁₂H₂₁N₅O₂S₂ Molecular Weight: 331,45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00865660 CAS-Nummer: 76963-41-2 Merck Index: 13,06694 |
Spezifikation Testergebnisse
Appearance | white to light yellow crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Melting point | 130°C to 134°C |
Loss on drying | ≤5 % (1 g, 105°C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...