Elvitegravir

Lieferant: Thermo Fisher Scientific

Synonyme: EVG, JTK-303/GS-9137, (S)-6-(3-Chloro-2-fluorobenzyl)-1-(1-hydroxy-3-methylbutan-2-yl)-7-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 6-[(3-chloro-2-fluorophenyl)methyl]-1-[(2S)-1-hydroxy-3-methylbutan-2-yl]-7-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

468510010 468512500
468510010.EA 978 EUR
468510010. 468512500.
Elvitegravir
Elvitegravir
Formel: C₂₃H₂₃ClFNO₅
Molecular Weight: 447.88 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11846135
CAS-Nummer: 697761-98-1

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR