Bicyclo[3.3.0]octane 97%

Lieferant: Thermo Fisher Scientific

Synonyme: Hexahydropentalene

468520010 468522500
468520010.EA 453 EUR
468520010. 468522500.
Bicyclo[3.3.0]octane 97%
Bicyclo[3.3.0]octane

Bestellen

Spezifikation Testergebnisse

Appearance clear colorless liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR