Pimobendan

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-(2-(4-Methoxyphenyl)-1H-benzo[d]imidazol-5-yl)-5-methyl-4,5-dihydropyridazin-3(2H)-one, 2-(2-Methyl-1-oxopropyl)isoindole-1,3-dione

468570010 468572500
468570010.EA 616 EUR
468570010. 468572500.
Pimobendan
Pimobendan
Formel: C₁₉H₁₈N₄O₂
Molecular Weight: 334.38 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00761648
CAS-Nummer: 74150-27-9
UN: 2811
ADR: 6.1 III
Merck Index: 13,07516

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Water ≤1 %
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR