Biapenem

Lieferant: Thermo Fisher Scientific

Synonyme: (4R,5S,6S)-3-((6,7-Dihydro-5H-pyrazolo[1,2-a][1,2,4]triazol-4-ium-6-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate

468660010 468662500
468660010.EA 1043 EUR
468660010. 468662500.
Biapenem
Biapenem
Formel: C₁₅H₁₈N₄O₄S
Molecular Weight: 350.39 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00864863
CAS-Nummer: 120410-24-4

Bestellen

Spezifikation Testergebnisse

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤2 %
Specific optical rotation -27° to -35° (20°C, 589 nm) (0.5% in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR