Etravirine

Lieferant: Thermo Fisher Scientific
Achtung

468721000 468725000
468721000.EA 102 EUR
468721000. 468725000.
Etravirine
Etravirine
Formel: C₂₀H₁₅BrN₆O
Lagertemperatur: Gefrierschrank
MDL: MFCD09837879
CAS-Nummer: 269055-15-4

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR