5'-Desoxy-5-fluorcytidin 95%
Lieferant: Thermo Scientific
Synonyme:
4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoropyrimidin-2(1H)-one
468740010
468740050
468740010.EA
29.3
EUR
468740010.
468740050.
5'-Desoxy-5-fluorcytidin 95%
5'-Desoxy-5-fluorcytidin
Formel:
C₉H₁₂FN₃O₄ Molecular Weight: 245,21 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07369278 CAS-Nummer: 66335-38-4 |
Spezifikation Testergebnisse
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Melting point | 210°C to 217°C |
Loss on drying | ≤0.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...