Oxibendazole

Lieferant: Thermo Fisher Scientific

Synonyme: (5-Propoxy-1H-benzimidazol-2-yl)carbamic acid methyl ester, Methyl (5-Propoxy-1H-benzo[d]imidazol-2-yl)carbamate

468810010 468810050
468810010.EA 67.9 EUR
468810010. 468810050.
Oxibendazole
Oxibendazole
Formel: C₁₂H₁₅N₃O₃
Molecular Weight: 249.27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00133728
CAS-Nummer: 20559-55-1
EINECS: 243-877-7
Merck Index: 13,07005

Bestellen

Spezifikation Testergebnisse

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR