Piroctone olamine 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine, 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2(1H)-one 2-aminoethanol salt

468820010 468820050
468820010.EA 44.7 EUR
468820010. 468820050.
Piroctone olamine 97%
Piroctone olamine
Formel: C₁₄H₂₃NO₂
Molecular Weight: 298,43 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01690792
CAS-Nummer: 68890-66-4
EINECS: 272-574-2
Merck Index: 13,07585

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Melting point 130°C to 138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR