Bevantolol hydrochloride
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-[(3,4-Dimethoxyphenethyl)amino]-3-(m-tolyloxy)propan-2-ol hydrochloride, 1-[[2-(3,4-Dimethoxyphenyl)ethyl]amino]-3-(3-methylphenoxy)-2-propanol hydrochloride
468870010
468872500
468870010.EA
376
EUR
468870010.
468872500.
Bevantolol hydrochloride
Bevantolol hydrochloride
Formel:
C₂₀H₂₇NO₄·HCl Molecular Weight: 381.90 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00941389 CAS-Nummer: 42864-78-8 Merck Index: 13,01194 |
Spezifikation Testergebnisse
Appearance | white to yellow to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 137°C to 140°C |
Water | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...