Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Bromfenac sodium

468920010 468922500
468920010.EA 437 EUR
468920010. 468922500.
Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate 98%
Sodium 2-(2-amino-3-(4-bromobenzoyl)phenyl)acetate
Formel: C₁₅H₁₁BrNNaO₃
Molecular Weight: 356,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00864341
CAS-Nummer: 91714-93-1
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance yellow to orange powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR