Canertinib dihydrochloride 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: CI-1033, PD-183805

468930010 468932500
468930010.EA 1434 EUR
468930010. 468932500.
Canertinib dihydrochloride 98%
Canertinib dihydrochloride
Formel: C₂₄H₂₅ClFN₅O₃·2HCl
Molecular Weight: 558,86 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09954112
CAS-Nummer: 289499-45-2

Bestellen

Spezifikation Testergebnisse

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Water ≤10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR