4-Bromisatin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Bromoindoline-2,3-dione
469060010
469060050
469060250
469060010.EA
25.1
EUR
469060010.
469060050.
469060250.
4-Bromisatin
4-Bromisatin
Formel:
C₈H₄BrNO₂ Molecular Weight: 226,0269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03618555 CAS-Nummer: 20780-72-7 |
Spezifikation Testergebnisse
Appearance | light yellow to brown crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Loss on drying | ≤2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...