Clevudine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-((2S,3R,4S,5S)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, 1-(2'-Deoxy-2'-fluoro-beta-L-arabinofuranosyl)-5-methyluracil
469120010
469122500
469120010.EA
590
EUR
469120010.
469122500.
Clevudine 97%
Clevudine
Formel:
C₁₀H₁₃FN₂O₅ Molecular Weight: 260.2200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00935785 CAS-Nummer: 163252-36-6 |
Spezifikation Testergebnisse
Appearance | white to light yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Loss on drying | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...