Oxfendazole 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl (6-(phenylsulphinyl)-1H-benzo[d]imidazol-2-yl)carbamate, Methyl [5-(Phenylsulphinyl)benzimidazol-2-yl]carbamate, [5-(Phenylsulphinyl)benzimidazol-2-yl]carbamic acid Methyl Ester
469210010
469210050
469210010.EA
24.2
EUR
469210010.
469210050.
Oxfendazole 98%
Oxfendazole
Formel:
C₁₅H₁₃N₃O₃S Molecular Weight: 315,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00801063 CAS-Nummer: 53716-50-0 EINECS: 258-714-5 Merck Index: 13,07004 |
Spezifikation Testergebnisse
Appearance | white to light yellow to light beige powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Loss on drying | ≤1 % (105°C, 2 h) |
Titration with HClO4 | 97.5 to 100.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...