9-Phenylacridin 98%

Lieferant: Thermo Fisher Scientific
Achtung

469430010 469430050 469430250
469430010.EA 23.2 EUR
469430010. 469430050. 469430250.
9-Phenylacridin 98%
9-Phenylacridine
Formel: C₁₉H₁₃N
Molecular Weight: 255.32 g/mol
Siedepunkt: 411°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00092562
CAS-Nummer: 602-56-2
EINECS: 210-020-3

Bestellen

Spezifikation Testergebnisse

Appearance light yellow to yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 183°C to 187°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR