Probucol 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4,4'-(Isopropylidendithio)bis[2,6-di-tert-butylphenol], 4,4'-(Propane-2,2-diylbis(sulphanediyl))bis(2,6-di-tert-butylphenol)

469490050 469490250
469490050.EA 41.8 EUR
469490050. 469490250.
Probucol 98%
Probucol
Formel: C₃₁H₄₈O₂S₂
Molecular Weight: 516.84 g/mol
Siedepunkt: 547°C (760mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00079281
CAS-Nummer: 23288-49-5
EINECS: 245-560-9
Merck Index: 13,07844

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 125°C to 128°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR