Piribedil 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-[4-(1,3-Benzodioxol-5-ylmethyl)-1-piperazinyl]pyrimidine, 2-(4-(Benzo[d][1,3]dioxol-5-ylmethyl)piperazin-1-yl)pyrimidine
469540010
469540050
469540250
469540010.EA
38.1
EUR
469540010.
469540050.
469540250.
Piribedil 98%
Piribedil
Formel:
C₁₆H₁₈N₄O₂ Molecular Weight: 298,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00868264 CAS-Nummer: 3605-01-4 EINECS: 222-764-6 Merck Index: 13,07576 |
Spezifikation Testergebnisse
Appearance | white to almost white crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 96°C to 100°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...