Morpholin-4-ylacetic acid 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
469580010
469580050
469580010.EA
31.2
EUR
469580010.
469580050.
Morpholin-4-ylacetic acid 95%
Morpholin-4-ylacetic acid
Formel:
C₆H₁₁NO₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD00504633 CAS-Nummer: 3235-69-6 |
Spezifikation Testergebnisse
Appearance | almost white to yellow crystalline powder |
Infrared spectrum | Conforms |
GC | ≥94 % |
Melting point | 160°C to 165°C |
Titration with NaOH | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...