3-Pyridineacetic acid 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(Pyridin-3-yl)acetic acid
469620010
469620050
469620010.EA
34.3
EUR
469620010.
469620050.
3-Pyridineacetic acid 95%
3-Pyridineacetic acid
Formel:
C₇H₇NO₂ Molecular Weight: 137,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066302 CAS-Nummer: 501-81-5 EINECS: 207-928-7 |
Spezifikation Testergebnisse
Appearance | white to beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Melting point | 143°C to 148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...