4-(Dimethylamino)butanoic acid 95%

Lieferant: Thermo Scientific
Achtung

469710010 469710050
469710010.EA 96.1 EUR
469710010. 469710050.
4-(Dimethylamino)butanoic acid 95%
4-(Dimethylamino)butanoic acid
Formel: C₆H₁₃NO₂
Molecular Weight: 131,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044628
CAS-Nummer: 693-11-8

Bestellen

Spezifikation Testergebnisse

Appearance white solid
Infrared spectrum Conforms
Titration with HClO4 94 to 106 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR