5-Chlorchinolin 97%

Lieferant: Thermo Scientific
Achtung

469740010 469740050
469740010.EA 44.9 EUR
469740010. 469740050.
5-Chlorchinolin 97%
5-Chloroquinoline
Formel: C₉H₆ClN
Molecular Weight: 163,61 g/mol
Flammpunkt: 263°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00239413
CAS-Nummer: 635-27-8

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
Melting point 28°C to 32°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR