Sulphadimethoxine 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Amino-N-(2,6-dimethoxypyrimidin-4-yl)benzenesulphonamide
469760050
469760250
469761000
469760050.EA
26.9
EUR
469760050.
469760250.
469761000.
Sulphadimethoxine 98%
Sulphadimethoxine
Formel:
C₁₂H₁₄N₄O₄S Molecular Weight: 310,33 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00057345 CAS-Nummer: 122-11-2 EINECS: 204-523-7 |
Spezifikation Testergebnisse
Appearance | white to light yellow to cream crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 195°C to 203°C |
Loss on drying | ≤0.5 % (1 g, 105°C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...