Tizanidine hydrochloride 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)benzo[c][1,2,5]thiadiazol-4-amine hydrochloride, Tizanidine HCl, 5-Chloro-4-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole Hydrochloride

470000050 470000250
470000050.EA 59.4 EUR
470000050. 470000250.
Tizanidine hydrochloride 98%
Tizanidine hydrochloride
Formel: C₉H₈ClN₅S
Molecular Weight: 290,17 g/mol
Siedepunkt: 391,2°C (760mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00798231
CAS-Nummer: 64461-82-1
Merck Index: 13,09561

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to light brown powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration Argentometric 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR