Rabeprazole sodium salt 98%
Lieferant: Thermo Fisher Scientific
470020010
470020050
470020010.EA
185
EUR
470020010.
470020050.
Rabeprazole sodium salt 98%
Rabeprazole sodium salt
Formel:
C₁₈H₂₁N₃O₃S Lagertemperatur: Gefrierschrank |
MDL:
MFCD02092688 CAS-Nummer: 117976-90-6 |
Spezifikation Testergebnisse
Appearance | white to yellow to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 145°C to 155°C |
Water | ≤10 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...