Thiazolidine-4-carboxylic acid 95%

Lieferant: Thermo Fisher Scientific
Achtung

470800010 470800050
470800010.EA 68.8 EUR
470800010. 470800050.
Thiazolidine-4-carboxylic acid 95%
Thiazolidine-4-carboxylic acid
Formel: C₄H₇NO₂S
Molecular Weight: 133.1689 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128958
CAS-Nummer: 444-27-9
EINECS: 207-146-6

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to brown powder or crystals
Infrared spectrum Conforms
HPLC ≥94 %
Titration with NaOH 94 to 106 % (On dry substance)
Water ≤10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR