1-Chlorethyl-isopropylcarbonat 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Carbonic Acid 1-Chloroethyl Isopropyl Ester, 1-Chloroethyl isopropyl carbonate
470930250
470931000
470930250.EA
29.9
EUR
470930250.
470931000.
1-Chlorethyl-isopropylcarbonat 98%
1-Chloroethylisopropyl carbonate
Formel:
C₆H₁₁ClO₃ Molecular Weight: 166,60 g/mol Siedepunkt: 164,3°C Dichte: 1,09 Flammpunkt: 55°C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038100 CAS-Nummer: 98298-66-9 UN: 2920 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | colorless to almost colorless liquid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Refractive index | 1.4110 to 1.4150 (20°C, 589 nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...