Nitazoxanid 98%
Lieferant: Thermo Scientific
Synonyme:
NTZ
471080010
471080050
471080250
471080010.EA
29.9
EUR
471080010.
471080050.
471080250.
Nitazoxanid 98%
Nitazoxanide
Formel:
C₁₂H₉N₃O₅S Molecular Weight: 307,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00416599 CAS-Nummer: 55981-09-4 EINECS: 259-931-8 Merck Index: 13,06595 |
Spezifikation Testergebnisse
Appearance | white to beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 198°C to 202°C |
Water | ≤10 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...