Trichostatin A 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: TSA, (R,2E,4E)-7-(4-(dimethylamino)phenyl)-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide, [R-(E,E)]-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide

471120050 471120250
471120050.EA 165 EUR
471120050. 471120250.
Trichostatin A 98%
Trichostatin A
Formel: C₁₇H₂₂N₂O₃
Molecular Weight: 302,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03848392
CAS-Nummer: 58880-19-6
Merck Index: 13,09722

Bestellen

Spezifikation Testergebnisse

Appearance light yellow to light pinkish to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 145°C to 150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR