2-Propylpyridine-4-carbothioamide 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Propyl-4-pyridinecarbothioamide, 2-Propylthioisonicotinamide, 2-Propylpyridine-4-carbothioamide, Prothionamide

471200050 471200250 471201000
471200050.EA 49.3 EUR
471200050. 471200250. 471201000.
2-Propylpyridine-4-carbothioamide 98%
2-Propylpyridine-4-carbothioamide
Formel: C₉H₁₂N₂S
Molecular Weight: 180,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00464119
CAS-Nummer: 14222-60-7
EINECS: 238-093-7
Merck Index: 13,07982

Bestellen

Spezifikation Testergebnisse

Appearance yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR