Phlorizin dihydrate 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(2,4-Dihydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)phenyl)-3-(4-hydroxyphenyl)propan-1-one dihydrate
471360010
471360050
471360010.EA
49
EUR
471360010.
471360050.
Phlorizin dihydrate 98%
Phlorizin dihydrate
Formel:
C₂₁H₂₈O₁₂ Molecular Weight: 472,43 g/mol Siedepunkt: 770°C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006591 CAS-Nummer: 7061-54-3 |
Spezifikation Testergebnisse
Appearance | light yellow to beige to light brown powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Water | 7 to 8.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...